R&D Systems
Manufacturer Code:AF3797
Catalog # AF3797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Smad3 Ser2-Ala230 Accession # P84022 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: hMAD-2; hMAD-2 hSMAD2 JV18 JV18-1 JV18-1SMAD mothers against DPP homolog 2 (Drosophila) MAD homolog 2 Mad protein homolog Mad mothers against decapentaplegic homolog 2 MADH2 MADH2MGC34440 MADR2 MADR2MGC22139 Mad-related protein 2 mother against DPP homolog 2 mothers against decapentaplegic homolog 2 Mothers against DPP homolog 2 Sma- and Mad-related protein 2 SMAD 2 SMAD family member 2MAD mothers against decapentaplegic homolog 2 (Drosophila) SMAD mothers against DPP homolog 2 Smad2 Smox; JV18-1; MAD homolog 2; Mad-related protein 2; mother against DPP homolog 2; Mothers against decapentaplegic homolog 2; Sma- and Mad-related protein 2; SMAD 2; SMAD family member 2; SMAD, mothers against DPP homolog 2
Gene Aliases: hMAD-2; hSMAD2; JV18; JV18-1; MADH2; MADR2; SMAD2
UniProt ID: (Human) Q15796
Entrez Gene ID: (Human) 4087
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.