R&D Systems
Manufacturer Code:IC7408T100UG
Catalog # IC7408T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 666633 |
Immunogen | <em>E. coli</em>-derived recombinant human Slug Met1-His268 Accession # O43623 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC10182 Neural crest transcription factor Slug Protein snail homolog 2 slug homolog zinc finger protein (chicken) SLUGH SLUGH1 SLUGzinc finger protein SNAI2 snail 2 snail homolog 2 (Drosophila) SNAIL2 WS2D zinc finger protein SNAI2; Neural crest transcription factor Slug; Protein snail homolog 2; slug (chicken homolog), zinc finger protein; snail family zinc finger 2; snail homolog 2; Zinc finger protein SNAI2
Gene Aliases: SLUG; SLUGH; SLUGH1; SNAI2; SNAIL2; WS2D
UniProt ID: (Human) O43623
Entrez Gene ID: (Human) 6591
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.