Invitrogen
Manufacturer Code:PA520289
Catalog # PIPA520289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide from near the center of human Slug. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neural crest transcription factor Slug; Protein snail homolog 2; slug (chicken homolog), zinc finger protein; SLUG SLUGH1 SNAIL2 WS2D neural crest transcription factor SLUG protein snail homolog 2 slug (chicken homolog) zinc finger protein zinc finger protein SNAI2; slug, chicken homolog; snail family zinc finger 2; snail homolog 2; Zinc finger protein SNAI2
Gene Aliases: SLUG; SLUGH; SLUGH1; SNAI2; SNAIL2; WS2D
UniProt ID: (Human) O43623, (Mouse) P97469
Entrez Gene ID: (Human) 6591, (Mouse) 20583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.