Novus Biologicals
Manufacturer Code:NBP176733
Catalog # NBP176733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 13 amino acid synthetic peptide from near the center of human Slug. The immunogen is located within amino acids 90 - 140 of Slug. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: MGC10182 Neural crest transcription factor Slug Protein snail homolog 2 slug homolog zinc finger protein (chicken) SLUGH SLUGzinc finger protein snail 2 snail homolog 2 (Drosophila) WS2D zinc finger protein SNAI2; Neural crest transcription factor Slug; Protein snail homolog 2; slug (chicken homolog), zinc finger protein; snail family zinc finger 2; snail homolog 2; Zinc finger protein SNAI2
Gene Aliases: SLUG; SLUGH; SLUGH1; SNAI2; SNAIL2; WS2D
UniProt ID: (Human) O43623
Entrez Gene ID: (Human) 6591
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.