Invitrogen
Manufacturer Code:PA520478
Catalog # PIPA520478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide from near the center of human Slitrk5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: leucine rich repeat containing 11; Leucine-rich repeat-containing protein 11; LRRC11 bA364G4.2 SLIT and NTRK-like protein 5 leucine rich repeat containing 11 leucine-rich repeat-containing protein 11 slit and trk like gene 5; SLIT and NTRK-like family, member 5; SLIT and NTRK-like protein 5; slit and trk like gene 5
Gene Aliases: 2610019D03Rik; bA364G4.2; KIAA0918; LRRC11; SLITRK5
UniProt ID: (Human) O94991, (Mouse) Q810B7
Entrez Gene ID: (Human) 26050, (Mouse) 75409, (Rat) 306152
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.