Invitrogen
Manufacturer Code:PA547519
Catalog # PIPA547519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Slit3 Ser27-His901 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MEGF5 SLIL2 KIAA0814 MEGF5Multiple epidermal growth factor-like domains protein 5 Multiple EGF-like domains protein 5 SLIL2 SLIL2FLJ10764 slit homolog 3 (Drosophila) slit homolog 3 protein SLIT1 slit2 Slit-3slit (Drosophila) homolog 3; Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; Multiple epidermal growth factor-like domains protein 5; slit homolog 3; Slit homolog 3 protein; Slit-3
Gene Aliases: b2b2362.1Clo; KIAA0814; MEGF5; SLIL2; Slit-3; SLIT1; slit2; SLIT3; UNQ691/PRO1336
UniProt ID: (Human) O95804, (Rat) O88280, (Mouse) Q9WVB4
Entrez Gene ID: (Human) 6586, (Rat) 83467, (Mouse) 20564
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.