Novus Biologicals
Manufacturer Code:NBP213350
Catalog # NBP213350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: CDNKNDSANACSAFKCHHGQCHISDQGEPYCLCQPGFSGEHCQQENPCLG QVVREVIRRQKGYASCATASKVPIMECRG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0814 MEGF5Multiple epidermal growth factor-like domains protein 5 Multiple EGF-like domains protein 5 SLIL2 SLIL2FLJ10764 slit homolog 3 (Drosophila) slit homolog 3 protein SLIT1 slit2 Slit-3slit (Drosophila) homolog 3; Multiple EGF-like domains protein 5; Multiple epidermal growth factor-like domains protein 5; slit homolog 3; Slit homolog 3 protein; Slit-3
Gene Aliases: KIAA0814; MEGF5; SLIL2; Slit-3; SLIT1; slit2; SLIT3; UNQ691/PRO1336
UniProt ID: (Human) O95804
Entrez Gene ID: (Human) 6586
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.