R&D Systems
Manufacturer Code:FAB5444R100UG
Catalog # FAB5444R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 710305 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2 Gln26-Gln900 Accession # Q9R1B9 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ14420 SLIL3 slit (Drosophila) homolog 2 slit homolog 2 (Drosophila) slit homolog 2 protein Slit-2SLIL3; Slit homolog 2 protein; Slit homolog 2 protein C-product; Slit homolog 2 protein N-product; Slit-2
Gene Aliases: SLIL3; Slit-2; SLIT2
UniProt ID: (Human) O94813
Entrez Gene ID: (Human) 9353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.