R&D Systems
Manufacturer Code:AF5444SP
Catalog # AF5444SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Slit2 Gln26-Gln900 Accession # Q9R1B9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ14420 SLIL3 slit (Drosophila) homolog 2 slit homolog 2 (Drosophila) slit homolog 2 protein Slit-2SLIL3; Slit homolog 2 protein; Slit homolog 2 protein C-product; Slit homolog 2 protein N-product; Slit-2
Gene Aliases: SLIL3; Slit-2; SLIT2
UniProt ID: (Human) O94813
Entrez Gene ID: (Human) 9353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.