Abnova
Manufacturer Code:PAB5908
Catalog # 89-131-997
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Slc9a3r1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3r1.
For Research Use Only
Protein Aliases: EBP-50; EBP50; Ezrin-radixin-moesin-binding phosphoprotein 50; MGC95776; Na(+)/H(+) exchange regulatory cofactor NHE-RF1; NHE-RF; NHERF-1; Regulatory cofactor of Na(+)/H(+) exchanger; Sodium-hydrogen exchanger regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1; Solute carrier family 9 isoform A3 regulatory factor 1
Gene Aliases: EBP-50; NHE-RF; Nherf; NHERF-1; NHERF1; Slc9a3r1
UniProt ID: (Mouse) P70441
Entrez Gene ID: (Mouse) 26941
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.