Abnova
Manufacturer Code:MAB6236
Catalog # 89-112-756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Slc6a2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of Slc6a2.
For Research Use Only
Protein Aliases: NE transporter; NE-T; NET; Norepinephrine transporter; Slc6a5; Sodium-dependent noradrenaline transporter; Solute carrier family 6 member 2
Gene Aliases: NE-T; NET; Slc6a2; Slc6a5
UniProt ID: (Mouse) O55192
Entrez Gene ID: (Mouse) 20538
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.