Abnova
Manufacturer Code:PAB8505
Catalog # 89-133-715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to rat Slc3a1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc3a1.
For Research Use Only
Protein Aliases: b(0,+)-type amino acid transport protein; D2; NAA-TR; Neutral and basic amino acid transport protein rBAT; Solute carrier family 3 member 1
Gene Aliases: Nbat; Slc3a1
UniProt ID: (Rat) Q64319
Entrez Gene ID: (Rat) 29484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.