Abnova
Manufacturer Code:MAB18254
Catalog # 89-338-179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S104-32 |
Immunogen | Recombinant protein corresponding to amino acids 1-63 of rat Slc38a1. |
Conjugate | ATTO488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Amino acid transporter A1; amino acid transporter system A1; Ata1; GlnT; Glutamine transporter; N-system amino acid transporter 2; rATA1; Sat1; Sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; System A amino acid transporter 1; System A transporter 2; System N amino acid transporter 1
Gene Aliases: Ata1; GlnT; Sa2; Sat1; Slc38a1; Snat1
UniProt ID: (Rat) Q9JM15
Entrez Gene ID: (Rat) 170567
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.