Invitrogen
Manufacturer Code:PA520554
Catalog # PIPA520554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the amino terminus of the human Slc35D1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 35 (UDP-GlcA/UDP-GalNAc transporter), member D1; solute carrier family 35 (UDP-glucuronic acid/UDP-N-acetylgalactosamine dual transporter), member D1; Solute carrier family 35 member D1; solute carrier family 35, member D1; UDP-galactose transporter-related 7; UDP-galactose transporter-related protein 7; UDP-GlcA/UDP-GalNAc transporter; UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter; UGTrel7; UGTREL7 UDP-GlcA/UDP-GalNAc transporter UDP-galactose transporter-related 7 UDP-galactose transporter-related protein 7 UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter solute carrier family 35 member D1
Gene Aliases: AI834976; C330011J09; KIAA0260; mKIAA0260; SLC35D1; UGTREL7
UniProt ID: (Human) Q9NTN3, (Mouse) A2AKQ0
Entrez Gene ID: (Human) 23169, (Rat) 298280, (Mouse) 242585
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.