Abnova
Manufacturer Code:PAB9637
Catalog # 89-134-291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus rat Slc32a1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc32a1.
For Research Use Only
Protein Aliases: GABA and glycine transporter; rGVAT; Solute carrier family 32 member 1; Vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat
Gene Aliases: Slc32a1; Vgat; Viaat
UniProt ID: (Rat) O35458
Entrez Gene ID: (Rat) 83612
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.