Abnova
Manufacturer Code:PAB14873
Catalog # 89-117-318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Slc25a19. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc25a19.
For Research Use Only
Protein Aliases: 2900089E13Rik; DNC; Mitochondrial thiamine pyrophosphate carrier; MUP1; solute carrier family 25 (mitochondrial deoxynucleotide carrier), member 19; Solute carrier family 25 member 19; TPC
Gene Aliases: 2900089E13Rik; DNC; MUP1; Slc25a19; TPC
UniProt ID: (Mouse) Q9DAM5
Entrez Gene ID: (Mouse) 67283
Molecular Function: RNA binding protein amino acid transporter calcium-binding protein calmodulin intracellular calcium-sensing protein mitochondrial carrier protein nucleic acid binding ribosomal protein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.