Abnova
Manufacturer Code:PAB8479
Catalog # 89-133-694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to rat Slc22a3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc22a3.
For Research Use Only
Protein Aliases: EMT; Oct3; Orct3; Organic cation transporter 3; Slca22a3; Solute carrier family 22 member 3; solute carrier family 22, member 3
Gene Aliases: EMT; Oct3; Orct3; Slc22a3; Slca22a3
UniProt ID: (Mouse) Q9WTW5
Entrez Gene ID: (Mouse) 20519
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.