Abnova
Manufacturer Code:MAB11522
Catalog # 89-109-672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S28-9 |
Immunogen | Recombinant fusion protien corresponding to amino acids 493-560 at C-terminus of rat Slc17a7. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant of Slc17a7.
For Research Use Only
Protein Aliases: BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGluT1
Gene Aliases: BNPI; Slc17a7; Vglut1
UniProt ID: (Rat) Q62634
Entrez Gene ID: (Rat) 116638
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.