Abnova
Manufacturer Code:PAB28836
Catalog # 89-130-359
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Slc14a2 C-terminus (amino acids 911-929). |
Conjugate | RPE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Rat Slc14a2.
For Research Use Only
Protein Aliases: plasma membrane urea transporter; Slc14a2T; solute carrier family 14 (urea transporter), member 2, variant 4; solute carrier family 14 (urea transporter), member 2T; Solute carrier family 14 member 2; solute carrier family 14, member 2; testName; testSymbol; urea transport protein; Urea transporter 2; Urea transporter, kidney; UrT1-C; UrT1-D
Gene Aliases: Slc14a2; Slc14a2_v4; Slc14a2T; testSymbol; UrT1-C; UrT1-D; Ut2
UniProt ID: (Rat) Q62668
Entrez Gene ID: (Rat) 54302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.