Abnova
Manufacturer Code:MAB18178
Catalog # 89-338-103
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S1-12 |
Immunogen | Recombinant protein corresponding to amino acids 932-1043 of rat Slc12a5. |
Conjugate | ATTO488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral potassium-chloride cotransporter 2; Furosemide-sensitive K-Cl cotransporter; K-Cl cotransporter 2; Kcc2; Neuronal K-Cl cotransporter; neuronal-specific K-Cl cotransporter; rKCC2; Solute carrier family 12 member 5; solute carrier family 12, member 5
Gene Aliases: Kcc2; Slc12a5
UniProt ID: (Rat) Q63633
Entrez Gene ID: (Rat) 171373
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.