Abnova
Manufacturer Code:PAB29037
Catalog # 89-130-550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 33-55 at N-terminus of rat Slc12a1. |
Conjugate | ATTO 390 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
For Research Use Only
Protein Aliases: bumetanide-sensitive sodium-(potassium)-chloride cotransporter 2; kidney-specific Na-K-Cl symporter; MGC93479; Nkcc2; solute carrier family 12 (sodium/potassium/chloride transporters), member 1; solute carrier family 12 member 1; Solute carrier family 12 member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter); Solute carrier family 12, member 1 (bumetanide-sensitive sodium-[potassium]-chloride cotransporter)
Gene Aliases: Nkcc2
Entrez Gene ID: (Rat) 25065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.