Invitrogen
Manufacturer Code:PA520307
Catalog # PIPA520307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide from near the amino terminus of human Ski. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Proto-oncogene c-Ski; Ski oncogene; ski oncoprotein; SKV C-ski proto-oncogene c-Ski ski oncogene ski oncoprotein; v-ski avian sarcoma viral oncogene homolog
Gene Aliases: SGS; SKI; SKV
UniProt ID: (Human) P12755
Entrez Gene ID: (Human) 6497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.