Invitrogen
Manufacturer Code:MA512542
Catalog # PIMA512542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C5.F8.C7 (alpha-Sr-1) |
Immunogen | N-terminal decapeptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: actin alpha 1; actin alpha skeletal muscle alpha skeletal muscle actin alpha-actin-1; actin, alpha 2, smooth muscle, aorta; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; alpha actin; alpha actin 2; Alpha-actin-1; Alpha-actin-2; alpha-cardiac actin; alpha-smooth muscle actin; Cell growth-inhibiting gene 46 protein; cytoplasmic beta-actin; nemaline myopathy type 3; smooth muscle alpha-actin; unnamed protein product
Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACTA; Acta-2; ACTA1; ACTA2; Acts; ACTSA; Actsk-1; ACTVS; alphaSMA; ASMA; CFTD; CFTD1; CFTDM; GIG46; MPFD; MYMY5; NEM1; NEM2; NEM3; SHPM; SMalphaA
UniProt ID: (Human) P68133, (Bovine) P68138, (Rabbit) P62740, (Bovine) P62739, (Human) P62736, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737
Entrez Gene ID: (Human) 58, (Bovine) 281592, (Rabbit) 100009506, (Sheep) 101107662, (Rabbit) 100009271, (Bovine) 515610, (Human) 59, (Sheep) 101121051, (Guinea pig) 100716933, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Guinea pig) 100722917
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.