Novus Biologicals
Manufacturer Code:NBP227205SS
Catalog # NBP227205SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 400-450 from human SIRT1 was used as the immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 75SirT1; EC 3.5.1 S. cerevisiae homolog) 1 sirtuin 1; hSIR2; hSIRT1; NAD-dependent protein deacetylase sirtuin-1; NAD-dependent protein deacylase sirtuin-1; Regulatory protein SIR2 homolog 1; SIR2-like protein 1; SirtT1 75 kDa fragment; sirtuin type 1
Gene Aliases: SIR2; SIR2alpha; SIR2L1; SIRT1
UniProt ID: (Human) Q96EB6
Entrez Gene ID: (Human) 23411
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.