Invitrogen
Manufacturer Code:PA517074
Catalog # PIPA517074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the carboxy terminus of mouse SirT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 75SirT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent deacetylase sirtuin-1 OTTHUMP00000198111 OTTHUMP00000198112 SIR2-like protein 1 SIR2alpha hSIR2 hSIRT1 sir2-like 1 sirtuin type 1; NAD-dependent protein deacetylase sirtuin-1; NAD-dependent protein deacylase sirtuin-1; Regulatory protein SIR2 homolog 1; Sir2; sir2-like 1; SIR2-like protein 1; SIR2alpha; SirtT1 75 kDa fragment; sirtuin 1 ((silent mating type information regulation 2, homolog) 1
Gene Aliases: AA673258; Sir2; Sir2a; Sir2alpha; SIR2L1; Sirt1
UniProt ID: (Mouse) Q923E4
Entrez Gene ID: (Mouse) 93759
Molecular Function: DNA binding protein chromatin/chromatin-binding protein deacetylase hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.