Novus Biologicals
Manufacturer Code:NBP159510
Catalog # NBP159510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SPPL2B(signal peptide peptidase-like 2B) The peptide sequence was selected from the N terminal of SPPL2B. Peptide sequence VARGNCTFYEKVRLAQGSGARGLLIVSRERLVPPGGNKTQYDEIGIPVAL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.23 EC 3.4.23.- IMP-4 IMP4MGC111084 Intramembrane protease 4 KIAA1532signal peptide peptidase-like 2B Presenilin-like protein 1 PSL1 SNPPL2B SPPL2b SPP-like 2B; IMP-4; Intramembrane protease 4; Presenilin homologous protein 4; Presenilin-like protein 1; PSH4; Signal peptide peptidase-like 2B; SPP-like 2B
Gene Aliases: IMP-4; IMP4; KIAA1532; PSH4; PSL1; SPPL2B
UniProt ID: (Human) Q8TCT7
Entrez Gene ID: (Human) 56928
Molecular Function: aspartic protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.