Novus Biologicals
Manufacturer Code:NBP17411120UL
Catalog # NB17419520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the middle region of H13. Immunizing peptide sequence IKLVFPQDLLEKGLEADNFAMLGLGDIVIPGIFIALLLRFDISLKKNTHT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ324O17.1 EC 3.4.23.- H13hIMP1 histocompatibility (minor) 13 IMP-1 IMP1MSTP086 Intramembrane protease 1 minor histocompatibility antigen 13 minor histocompatibility antigen H13 Presenilin-like protein 3 PSENL3 PSL3IMPAS Signal peptide peptidase signal peptide peptidase beta SPPIMPAS-1; histocompatibility (minor) 13; histocompatibility 13; IMP-1; Intramembrane protease 1; minor histocompatibility antigen 13; Minor histocompatibility antigen H13; Presenilin-like protein 3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1
Gene Aliases: H13; HM13; IMP1; IMPAS; IMPAS-1; MSTP086; PSENL3; PSL3; SPP; SPPL1
UniProt ID: (Human) Q8TCT9
Entrez Gene ID: (Human) 81502
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.