Invitrogen
Manufacturer Code:PA512327
Catalog # PIPA512327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 118-147 amino acids from the central region of human OPRS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aging-associated gene 8 protein; opioid receptor, sigma 1; SIG-1R; sigma 1 receptor variant SR-1D; Sigma 1-type opioid receptor; Sigma 1-type opioid receptor Sigma1-receptor Sigma1R SIG-1R hSigmaR1 SR31747-binding protein SR-BP Aging-associated gene 8 protein OPRS1; Sigma non-opioid intracellular receptor 1; Sigma1-receptor; SR-BP; SR31747 binding protein 1; SR31747-binding protein
Gene Aliases: AAG8; ALS16; DSMA2; hSigmaR1; OPRS1; SIG-1R; sigma1R; SIGMAR1; SR-BP; SR-BP1; SRBP
UniProt ID: (Human) Q99720, (Mouse) O55242
Entrez Gene ID: (Human) 10280, (Mouse) 18391
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.