Novus Biologicals
Manufacturer Code:NBP219628
Catalog # NBP219628
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human SIGMAR1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aging-associated gene 8 protein; Aging-associated gene 8 protein FLJ25585 hSigmaR1 OPRS1 SIG-1R sigma 1 Sigma 1-type opioid receptor sigma non-opioid intracellular receptor 1 sigma1 receptor sigma1R sigma1-receptor SR31747 binding protein 1 SR31747-binding protein SR-BP SR-BP1 SRBPMGC3851 type I sigma receptor; SIG-1R; Sigma 1-type opioid receptor; Sigma non-opioid intracellular receptor 1; SR-BP; SR31747 binding protein 1; SR31747-binding protein
Gene Aliases: AAG8; ALS16; DSMA2; hSigmaR1; OPRS1; SIG-1R; sigma1R; SIGMAR1; SR-BP; SR-BP1; SRBP
UniProt ID: (Human) Q99720
Entrez Gene ID: (Human) 10280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.