Novus Biologicals
Manufacturer Code:NBP147969
Catalog # NBP147969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D9 |
Immunogen | Full length human recombinant protein of human SIGLEC9 (NP_055256) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD329; CD329 CDw329 FOAP-9 OBBP-LIKE Protein FOAP-9 sialic acid binding Ig-like lectin 9 sialic acid-binding Ig-like lectin 9 siglec-9; CDw329; Protein FOAP-9; sialic acid binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin 9; Siglec-9
Gene Aliases: CD329; CDw329; FOAP-9; OBBP-LIKE; siglec-9; SIGLEC9; UNQ668/PRO1302
UniProt ID: (Human) Q9Y336
Entrez Gene ID: (Human) 27180
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.