R&D Systems
Manufacturer Code:FAB1139N025
Catalog # FAB1139N025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 191240 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Siglec-9 Gln18-Gly348 (predicted) Accession # Q9Y336 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: CD329; CD329 CDw329 FOAP-9 OBBP-Like Protein FOAP-9 sialic acid binding Ig-like lectin 9 sialic acid-binding Ig-like lectin 9 siglec-9 Siglec9; CDw329; Protein FOAP-9; sialic acid binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin 9; Siglec-9
Gene Aliases: CD329; CDw329; FOAP-9; OBBP-LIKE; siglec-9; SIGLEC9; UNQ668/PRO1302
UniProt ID: (Human) Q9Y336
Entrez Gene ID: (Human) 27180
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
membrane-bound signaling molecule
myelin protein
receptor
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.