Novus Biologicals
Manufacturer Code:NBP237328
Catalog # NBP237328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A3A11 |
Immunogen | Purified recombinant fragment of human CD22 (AA: 621-725) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B-cell receptor CD22; B-cell receptor CD22 BL-CAM B-lymphocyte cell adhesion molecule CD22 antigenMGC130020 CD22 molecule sialic acid binding Ig-like lectin 2 Sialic acid-binding Ig-like lectin 2 SIGLEC-2 SIGLEC2FLJ22814 T-cell surface antigen Leu-14; B-lymphocyte cell adhesion molecule; BL-CAM; CD22; CD22 antigen; sialic acid binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2; Siglec-2; T-cell surface antigen Leu-14
Gene Aliases: CD22; SIGLEC-2; SIGLEC2
UniProt ID: (Human) P20273
Entrez Gene ID: (Human) 933
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule membrane-bound signaling molecule myelin protein receptor signaling molecule structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.