Novus Biologicals
Manufacturer Code:NBP294757100UL
Catalog # NB024033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SLC17A5 (NP_036566.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AST; ASTSodium/sialic acid cotransporter FLJ22227 FLJ23268 ISSD Membrane glycoprotein HP59 NSD SD sialic acid storage disease SIALIN SIASD SLDSolute carrier family 17 member 5 solute carrier family 17 (anion/sugar transporter) member 5 solute carrier family 17 member 5; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; Membrane glycoprotein HP59; sialic acid storage disease; Sialin; sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; solute carrier family 17 (anion/sugar transporter), member 5; Solute carrier family 17 member 5; Vesicular H(+)/Aspartate-glutamate cotransporter
Gene Aliases: AST; ISSD; NSD; SD; SIALIN; SIASD; SLC17A5; SLD
UniProt ID: (Human) Q9NRA2
Entrez Gene ID: (Human) 26503
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.