Invitrogen
Manufacturer Code:PA517167
Catalog # PIPA517167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide surrounding Lys450 of human shootin1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Shootin-1; shootin-1 shootin1; Shootin1
Gene Aliases: KIAA1598; mKIAA1598; Shootin1; SHTN1
UniProt ID: (Mouse) Q8K2Q9, (Rat) A0MZ67, (Human) A0MZ66
Entrez Gene ID: (Mouse) 71653, (Rat) 292139, (Human) 57698
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.