Proteintech
Manufacturer Code:662971IG150UL
Catalog # 50-173-6729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A1A4 |
Immunogen | Sestrin 2 Fusion Protein Ag20638 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HI95 SES2 SESN2 SEST2 sestrin 2 Sestrin2; hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; Sestrin-2
Gene Aliases: HI95; RGD1566319; SES2; SESN2; SEST2
UniProt ID: (Human) P58004
Entrez Gene ID: (Pig) 100620966, (Human) 83667, (Rat) 502988
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.