Proteintech
Manufacturer Code:107951AP150UL
Catalog # 50-556-521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sestrin 2 Fusion Protein Ag1247 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HI95 SES2 SESN2 SEST2 Sestrin2 Sestrin-2; hypoxia induced gene 95; hypoxia-induced; Hypoxia-induced gene; Sestrin-2
Gene Aliases: HI95; RGD1566319; SES2; SESN2; SEST2
UniProt ID: (Human) P58004, (Mouse) P58043
Entrez Gene ID: (Human) 83667, (Rat) 502988, (Mouse) 230784
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.