Invitrogen
Manufacturer Code:PA566705
Catalog # PIPA566705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Seryl-tRNA synthetase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: serine tRNA ligase 1, cytoplasmic; Serine--tRNA ligase, cytoplasmic; SerRS; SERRS SERS Seryl-tRNA Ser/Sec synthetase serine tRNA ligase 1 cytoplasmic serine--tRNA ligase cytoplasmic seryl-tRNA synthetase cytoplasmic seryl-tRNA(Ser/Sec) synthetase; Seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: SARS; SARS1; SERRS; SERS
UniProt ID: (Human) P49591
Entrez Gene ID: (Human) 6301
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.