Invitrogen
Manufacturer Code:PA553521
Catalog # PIPA553521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Seryl-tRNA synthetase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: serine tRNA ligase 1, cytoplasmic; Serine--tRNA ligase, cytoplasmic; SerRS; SERRS SERS Seryl-tRNA Ser/Sec synthetase serine tRNA ligase 1 cytoplasmic serine--tRNA ligase cytoplasmic seryl-tRNA synthetase cytoplasmic seryl-tRNA(Ser/Sec) synthetase; seryl-aminoacyl-tRNA synthetase 1; Seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: SARS; SARS1; SERRS; SERS; Strs
UniProt ID: (Human) P49591, (Mouse) P26638, (Rat) Q6P799
Entrez Gene ID: (Human) 6301, (Mouse) 20226, (Rat) 266975
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.