Invitrogen
Manufacturer Code:MA523975
Catalog # PIMA523975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 273912 |
Immunogen | Mouse myeloma cell line NS0-derived recombit rat Pentraxin2/SAP Gln21-Ser228 (predicted) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PTX2 SAP 9.5S alpha-1-glycoprotein pentaxin-related serum amyloid P-component; SAP; Serum amyloid P-component
Gene Aliases: Apcs; Ptx2; Sap
UniProt ID: (Rat) P23680
Entrez Gene ID: (Rat) 29339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.