Invitrogen
Manufacturer Code:PA578789
Catalog # PIPA578789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse Serum Amyloid P recombit protein (Position: Q21-D224). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APCS amyloid P-component serum protein; SAP; Serum amyloid P-component; Serum amyloid P-component SAP Apcs Ptx2 Sap
Gene Aliases: Apcs; Ptx2; Sap
UniProt ID: (Mouse) P12246
Entrez Gene ID: (Mouse) 20219
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.