R&D Systems
Manufacturer Code:MAB30195100
Catalog # MAB30195100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 902770 |
Immunogen | <em>E. coli</em>-derived recombinant human Serum Amyloid A1 Arg19-Tyr133 Accession # P0DJI8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Amyloid fibril protein AA; Amyloid protein A; SAA; serum amyloid A protein; Serum amyloid A-1 protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); tumor protein p53 inducible protein 4
Gene Aliases: PIG4; SAA; SAA1; SAA2; TP53I4
UniProt ID: (Human) P0DJI8
Entrez Gene ID: (Human) 6288
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.