Abnova
Manufacturer Code:MAB3804
Catalog # 89-111-602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MA32K3 |
Immunogen | Recombinant protein corresponding to rat Serpine1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant Serpine1.
For Research Use Only
Protein Aliases: Endothelial plasminogen activator inhibitor; PAI; PAI-1; Pai1; PAI1A; Pai1aa; Planh; Plasminogen activator inhibitor 1; RATPAI1A; serine (or cysteine) peptidase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; serine (or cysteine) proteinase inhibitor, clade E, member 1; serine (or cysteine) proteinase inhibitor, member 1; Serpin E1
Gene Aliases: Pai1; PAI1A; Pai1aa; Planh; Planh1; RATPAI1A; Serpine1
UniProt ID: (Rat) P20961
Entrez Gene ID: (Rat) 24617
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.