Abnova
Manufacturer Code:MAB3808
Catalog # 89-111-606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H14H7 |
Immunogen | Recombinant protein corresponding to mouse Serpine1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant Serpine1.
For Research Use Only
Protein Aliases: endothelial plasminogen activator inhibitor; nexin, plasminogen activator inhibitor type 1; PAI; PAI-1; PAI1; Planh1; plasminogen activator inhibitor 1; plasminogen activator inhibitor, type I; Plasminogen activator inhibitor-1 precursor (PAI-1) (Endothelial plasminogen activator inhibitor) (PAI); serine (or cysteine) proteinase inhibitor, clade E, member 1; serpin E1
Gene Aliases: PAI-1; PAI1; Planh1
Entrez Gene ID: (Mouse) 18787
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.