Novus Biologicals
Manufacturer Code:NB10058965
Catalog # NB10058965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-PKFKLQEDYDME corresponding to internal region according to NP_004146.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CAP-3; CAP-3 CAP3PI-9 Cytoplasmic antiproteinase 3 Peptidase inhibitor 9 PI9protease inhibitor 9 (ovalbumin type) serine (or cysteine) proteinase inhibitor clade B (ovalbumin) member 9 serpin B9 serpin peptidase inhibitor clade B (ovalbumin) member 9 serpin peptidase inhibitor clade B member 9; Cytoplasmic antiproteinase 3; Peptidase inhibitor 9; PI-9; protease inhibitor 9 (ovalbumin type); serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 9; Serpin B9; serpin peptidase inhibitor, clade B (ovalbumin), member 9; serpin peptidase inhibitor, clade B, member 9; testicular tissue protein Li 180
Gene Aliases: CAP-3; CAP3; PI-9; PI9; SERPINB9
UniProt ID: (Human) P50453
Entrez Gene ID: (Human) 5272
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.