R&D Systems
Manufacturer Code:AF1177SP
Catalog # AF1177SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Serpin F1/PEDF Asp44-Pro418 Accession # P36955 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 35 protein; Cell proliferation-inducing gene 35 protein EPC-1 EPC-1PIG35 PEDF PEDFpigment epithelium-derived factor pigment epithelium derived factor) member 1 proliferation-inducing protein 35 serine (or cysteine) proteinase inhibitor clade F (alpha-2 antiplasmin Serpin F1 serpin peptidase inhibitor clade F (alpha-2 antiplasmin pigment epitheliumderived factor) member 1; EPC-1; PEDF; Pigment epithelium-derived factor; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; Serpin F1; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; sserpin family F member 1; testis tissue sperm-binding protein Li 70n
Gene Aliases: EPC-1; OI12; OI6; PEDF; PIG35; SERPINF1
UniProt ID: (Human) P36955
Entrez Gene ID: (Human) 5176
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.