Novus Biologicals
Manufacturer Code:NBP289393V
Catalog # NB066710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Serpin E1/PAI-1 (NP_000593.1 Met 1-Pro 402). |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Endothelial plasminogen activator inhibitor; Endothelial plasminogen activator inhibitor PAI1PAI-1 PAISerpin E1 PLANH1plasminogen activator inhibitor 1 serine (or cysteine) proteinase inhibitor clade E (nexin plasminogenactivator inhibitor type 1) member 1 serpin peptidase inhibitor clade E (nexin plasminogen activator inhibitortype 1) member 1; PAI; Plasminogen activator inhibitor 1; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; Serpin E1; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1
Gene Aliases: PAI; PAI-1; PAI1; PLANH1; SERPINE1
UniProt ID: (Human) P05121
Entrez Gene ID: (Human) 5054
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.