Novus Biologicals
Manufacturer Code:NBP213298
Catalog # NBP213298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: RLFHKSDGSTVSVPMMAQTNKFNYTEFTTPDGHYYDILELPYHGDTLSMF IAAPYEKEVPLSALTNILSAQLISHWKGNMTRLPRLLVLPKFSLETEVDL RKPLENLGMTDMFRQFQADF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Endothelial plasminogen activator inhibitor; Endothelial plasminogen activator inhibitor PAI1PAI-1 PAISerpin E1 PLANH1plasminogen activator inhibitor 1 serine (or cysteine) proteinase inhibitor clade E (nexin plasminogenactivator inhibitor type 1) member 1 serpin peptidase inhibitor clade E (nexin plasminogen activator inhibitortype 1) member 1; PAI; Plasminogen activator inhibitor 1; serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1; Serpin E1; serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1
Gene Aliases: PAI; PAI-1; PAI1; PLANH1; SERPINE1
UniProt ID: (Human) P05121
Entrez Gene ID: (Human) 5054
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.