Novus Biologicals
Manufacturer Code:NBP290150AF350
Catalog # NB070362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 309 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Serpin B6 (AAB30320.1 Asp 2-Pro 376). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CAP; CAPMGC111370 Cytoplasmic antiproteinase deafness autosomal recessive 91 DFNB91 Peptidase inhibitor 6 PI-6 PI6MSTP057 Placental thrombin inhibitor protease inhibitor 6 (placental thrombin inhibitor) PTIDKFZp686I04222 serine (or cysteine) proteinase inhibitor clade B (ovalbumin) member 6 serpin B6 serpin peptidase inhibitor clade B (ovalbumin) member 6 SPI3; Cytoplasmic antiproteinase; Peptidase inhibitor 6; PI-6; Placental thrombin inhibitor; protease inhibitor 6 (placental thrombin inhibitor); serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 6; Serpin B6; serpin peptidase inhibitor, clade B (ovalbumin), member 6
Gene Aliases: CAP; DFNB91; MSTP057; PI-6; PI6; PTI; SERPINB6; SPI3
UniProt ID: (Human) P35237
Entrez Gene ID: (Human) 5269
Molecular Function: enzyme modulator protease inhibitor serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.