Novus Biologicals
Manufacturer Code:NBP245788
Catalog # NBP245788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A12 |
Immunogen | Full length human recombinant protein of human SERPINB3(NP_008850) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HsT1196 Protein T4-A SCC SCCA SCCA1SCCA-1 SCCA-PD serine (or cysteine) proteinase inhibitor clade B (ovalbumin) member 3 serpin B3 serpin peptidase inhibitor clade B (ovalbumin) member 3 Squamous cell carcinoma antigen 1 T4-A; Protein T4-A; SCCA-1; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 3; Serpin B3; serpin peptidase inhibitor, clade B (ovalbumin), member 3; Squamous cell carcinoma antigen 1
Gene Aliases: HsT1196; SCC; SCCA; SCCA-1; SCCA-PD; SCCA1; SERPINB3; SSCA1; T4-A
UniProt ID: (Human) P29508
Entrez Gene ID: (Human) 6317
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.