R&D Systems
Manufacturer Code:MAB6966
Catalog # MAB6966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 754438 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Serpin A8/Angiotensinogen Asp25-Val477 Accession # P11859 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AGT alpha-1 antiproteinase antitrypsin angiotensin II Angiotensinogen angiotensinogen (serpin peptidase inhibitor clade A member 8) ANHU Aogen FLJ92595 FLJ97926 PAT pre-angiotensinogen serine (or cysteine) proteinase inhibitor Serpin A8 SERPINA8angiotensin I; alpha-1 antiproteinase, antitrypsin; Ang I; Ang II; Ang III; Ang IV; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; Angiotensin II; Angiotensin III; Angiotensin IV; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (serpin peptidase inhibitor, clade A, member 8); Des-Asp[1]-angiotensin II; pre-angiotensinogen; serine (or cysteine) proteinase inhibitor; Serpin A8; serpin peptidase inhibitor, clade A, member 8
Gene Aliases: AGT; ANHU; SERPINA8
UniProt ID: (Human) P01019
Entrez Gene ID: (Human) 183
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.